Clean-Ceil™ Fan Filter Modules, Microzone
Proveedor: Microzone
Clean-Ceil™
FFM-2-4-A
35995-762EA 43000.8 MXN
35995-762
Clean-Ceil™ Fan Filter Modules, Microzone
Habitaciones prefabricadas
Ideal for use in cleanroom and sterile suite ceiling systems
These terminal fan filter HEPA units may be incorporated into existing facilities as a retrofit to upgrade cleanliness levels, or may be integrated into the design of new cleanroom installations. The energy-efficient Clean-Ceil™ filter is also ideal for use in modular or portable cleanrooms. Standard filter module comes complete with removable, washable, 40% efficient ASHRAE-rated prefilter and solid-state speed control.