Samco™ Disposable Transfer Pipettes, Graduated, Thermo Scientific
Proveedor: Thermo Scientific
222-1S 225-20S 282-1S 225 274 212-20S 222-10S 274-1S 728NL 225-1S 282 222-20S 282-20S 274-1S 282 222 212-1S 718 212
73990-088CS 16364.04 MXN
73990-094 14670-307 490014-718 73990-096 73990-088 14670-201 14670-366 14670-200 14670-420 73990-036 14670-364 73990-026 490014-716 14670-205 75838-704 75838-706 14670-282 14670-363 14670-250
Samco™ Disposable Transfer Pipettes, Graduated, Thermo Scientific
Pipetas Pipetas de transferencia (Pasteur)
Graduated polyethylene transfer pipettes are excellent for sampling of serum, plasma, blood, urine, and other biological fluids, especially where more accurate volume measurements are required.
Sterile models are specially packaged for batch-testing applications in microbiology and bacteriology labs. Nonsterile models are suitable for single, specialized research type testing. Stem narrows down to a fine tip.