VWR® Disposable Transfer Pipettes, Graduated
Proveedor: VWR
VWR®
P122 16001-188 414004-013 414004-025 16001-182 414004-015 414004-024 414004-040 414004-043 16001-176 414004-045 414004-014 414004-035 414004-044 414004-034 16001-172 414004-017 414004-016 16001-180
414004-017CS 3313.46 MXN
76004-132 414004-017 414004-016 414004-015 16001-188 414004-025 414004-014 414004-013 414004-024 414004-035 414004-045 414004-034 414004-044 414004-043 414004-040 16001-176 16001-180 16001-182 16001-172
VWR® Disposable Transfer Pipettes, Graduated
Pipetas Pipetas de transferencia (Pasteur)
VWR® Disposable Transfer Pipettes are ideal for quick and safe fluid transfer.
  • Will not shatter or break
  • Can be used in liquid nitrogen
  • Nontoxic and inert
  • No bulb to insert or remove
  • Uniform drop size
These unbreakable all-in-one graduated transfer pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from see-through low density polyethylene, pipettes are inert to biological fluids and most acids, and can be sealed and refrigerated. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Ideal for quick, safe transfer of fluids. Transfer pipettes can be gas sterilized. Manufactured from LDPE, they can be sealed, refrigerated, and gas sterilized.