VWR® Rubber Bulbs
Proveedor: VWR
VWR®
20153 20155
15001-364PK 137.32 MXN
15001-364 15001-362
VWR® Rubber Bulbs
Pipetas Bombas de pipeteadores
Made of durable natural rubber, bulbs are suitable for medicine droppers and small dropping pipettes.
  • Durable
  • Natural rubber construction
  • Heavy-walled design
  • Cylindrical
Neck is grooved to take flat flange of pipettes. Bulbs fit pipettes 6 to 8 mm ext. Ø.