SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Proveedor: SP Industries
LG-9335-120 LG-9335-116 LG-9335-112 LG-9335-110 LG-9335-118
14212-502EA 419.27 MXN
14212-244 14212-246 14212-502 14211-580 14212-506
SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Pipetas Pipetas serológicas
Calibrated 'to deliver' (TD) at 20°C, these borosilicate glass pipettes have a permanently colored amber scale with the zero line at the top
  • Reference: ASTM E 1044, Lab Grade
The small amount remaining in the tip after completion of free delivery is blown out and added to the main delivery volume (ground band indicator at top). The 5,10, and 25 ml pipettes have openings of reduced diameter.
Precaución: NOTE: Do not pipette by mouth. A mechanical device is suggested.