VWR® Disposable Serological Pipettes, Glass
Proveedor: VWR
VWR®
VW72105-21100 VW72105-5110 VW72100-21100 VW72102-11100 VW72120-21100 VW72120-11100 VW72100-5110 VW72105-10110 VW72102-21100 VW72100-1110 VW72102-5110 VW72120-1110 VW72100-10110 VW72105-1110 VW72120-5110 VW72105-11100 VW72100-11100 VW72102-10110 VW72120-10110
93000-706CS 6764.65 MXN
93000-706 93000-704 93000-702 93000-678 93000-700 93000-690 93000-680 93000-676 93000-742 93000-698 93000-688 93000-696 93000-674 93000-686 93000-694 93000-672 93000-684 93000-692 93000-682
VWR® Disposable Serological Pipettes, Glass
Pipetas Pipetas serológicas
VWR® Disposable Serological Pipettes are available in sterile or nonsterile formats, with a variety of packaging options to choose from.
  • Uniform pulled tip, calibrated to deliver
  • Borosilicate glass construction
  • Negative graduations increase versatility
  • Available in sterile or nonsterile formats
These pipettes have a uniform pulled tip and are calibrated to deliver to the tip, with two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered. Negative graduations increase the versatility of the pipet. Pipettes are available sterile and plugged or in bulk packs of nonsterile, non-plugged pipettes. Nonsterile, non-plugged pipettes feature a blue scale. Made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. Individually wrapped, multi-pack, and canister pack pipettes are also designed from ASTM Specification E714.
Envase: Individually wrapped pipettes are packaged in color-coded paper/plastic wraps and overpacked in convenient dispensing shelf packs for laboratory benches. Multi-pack pipettes come in color-coded bags of 25 pipettes (1 ml and 2 ml sizes) or 10 pipettes (5 ml and 10 ml sizes). Canister pack pipettes are packaged in a reclosable pack of 50 pieces each sealed in a plastic bag. Bulk pack pipettes are sealed in plastic bags and overpacked in shelf packs.