E.Z.N.A.® RNA Isolation Kits, Omega Bio-Tek
Proveedor: Omega Bio-Tek
E.Z.N.A®
R6934-02 R6950-00 R6814-00 R6831-01 R6840-01 R6840-00 R6831-02 R6664-02 R6664-00 R6934-01 R6840-00 R6664-00 R6950-01 R6814-00 R6814-01 R6814-01 R6950-00 R6934-00 R6950-01 R6831-02 R6934-00 R6934-01 R6934-02 R6831-01 R6831-00 R6664-02 R6840-01 R6831-00
101319-236 101319-358 101319-216 CA101319-232 CA101319-234 CA101319-236 CA101319-358 CA101319-216 101319-232 101319-234 101319-224 CA101319-380 CA101319-260 CA101319-382 CA101319-262 CA101414-872 CA101319-264 CA101319-266 CA101319-244 CA101319-224 101319-380 101319-260 101319-382 101319-262 101414-872 101319-264 101319-244 101319-266
E.Z.N.A.® RNA Isolation Kits, Omega Bio-Tek
Reactivos de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
Isolated RNA can be used for Northern blotting, RT-PCR, nuclease protection assays, in vitro translation, and microarray analysis
MicroElute® kit 101319-234 provides a rapid and easy method for the isolation of up to 50µg of total RNA from small amounts of cultured eukaryotic cells, tissues such as laser dissected samples (LDS), or fine needle aspirates (FNA).
RNAsafer™ stabilizer reagent rapidly lyses cells and tissues to inactivate RNases at room temperature.
SQ RNA/DNA/protein isolation kits quickly, reliably, and simultaneously extract DNA, RNA, and proteins from a single sample without organic extractions.
Miniprep kits that use HiBind™ spin columns use V-spin columns which feature an attached cap and a standard luer tip at the bottom for easy, leak-free connection to a vacuum manifold or luer hub needle. Midi and maxi columns feature a standard vacuum leuer for faster processing. These columns can be used for both centrifugation and vacuum protocols.
Información para pedidos: Solution-based, silica column, and magnetic bead-based kits are available; contact your Avantor representative for more information.