Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Proveedor: Thermo Scientific
Zeba™
89894 87773 89891 87770 89893 87769 89882 87765 87768 89883 89893 89878 89889 89890 89890 89877 89892 89891 89892 87766 87767 87764 89882 87767 87771 89889 87768 87765 87770 87769 89877 89878 89894 89883
PI89890EA 10668 MXN
CAPI89878 CAPI89877 PI89889 PI89882 PI87766 PI87765 PI89883 PI87764 CAPI89883 CAPI89882 PI89890 PI89891 PI89892 CAPI87765 CAPI89889 CAPI87767 PI89877 CAPI87768 PI89878 PI87770 PI89893 CAPI87769 PI89894 CAPI89892 PI87769 CAPI89891 CAPI89894 CAPI87770 PI87768 CAPI89893 PI87767 CAPI87771 CAPI87773 CAPI89890
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Filtros Filtros centrífugos
The Zeba™ spin desalting columns are made of low protein binding polypropylene and are compatible with a wide range of standard laboratory instruments and consumables.
  • Higher protein recovery
  • Minimal sample dilution
  • No screening fractions for protein or waiting for protein to emerge by gravity-flow
  • Easy-to-use with no cumbersome column preparation or equilibration
The Zeba 7K MWCO is recommended for processing proteins >7 kDa, and the Zeba 40K MWCO for processing proteins >40 kDa.
They provide >95% retention of salts and other small molecules.
Precaución: Zeba™ columns are designed for research use only. Not for use in diagnostic procedures. Store at 4˚C.