VWR® Bi-Metallic Surface Temperature Thermometers
61320-1611 61320-1411 61320-0611
98000-258 98000-260 98000-232
VWR® Bi-Metallic Surface Temperature Thermometers
Surface temperature bi-metallic thermometers feature bi-metallic coil movement and a aluminum dial ideal for pipes, griddles and hot plates.
  • Accurate to within 2.5°
These surface thermometers are ideal for measuring temperatures in pipes, griddles, and hot plates.
Certificaciones: Tested and inspected by Avantors's USA quality team, these thermometers include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy. Should an individual Thermometer Calibration Report be required, custom calibration services are available.