Short-Term Detector Tubes, Draeger
Proveedor: Dräger
CH29101 CH21101 8101701 6733031 CH21101 6733181 6728531 6718401 8103261 6724001 8101521 8101321 8101781 8101621 8101831 8101721 8101721 6728491 8103261 6724001 8101641 CH23101 CH25903 8101991 6733141 CH31701 6730201 CH29801 8101211 6728531 6730201 6728161 6728161 CH28301 CH31201 CH19601 6728541 6728781 6723301 6724501 8101671 8101871 6733161 8101341 8101961 CH28101 8101341 CH24201 8101621 6733141 6728651 CH28301 8101661 CH28101 8101691 6728491 6728371 8101461 CH28201 8101501 CH21001 8103631 6728311 CH31001 CH25803 8101501 6728821 8101611 8101531 8101461 8101991 6733031 6728541 8101661 8101921 6728461 6728041 CH25803 8101781 CH26303 CH26303 8101831 8101961 6733161 CH28201 CH19601 8101611 CH20001 CH31001 6728781 8101211 6728461 CH29101 8101521 6728981 CH24201 6728371 CH31701 CH27301 CH21001 6728041 CH28401 8101531 CH27301 8101871 6723301 6728981 8101671 CH20001 6733181 CH29801 CH25903 8101321 CH30701 8101691 CH23101 6724501 8101641
13453-350PK 6215.76 MXN
13453-108 CA13453-118 13453-226 13453-228 13453-348 CA13453-114 CA13453-232 13453-222 CA13453-112 CA13453-198 CA13453-350 13453-220 13453-340 34331-688 CA13453-402 13453-118 CA13453-366 CA13453-400 CA13453-246 CA13453-122 13453-112 CA13453-364 13453-232 13453-114 13453-234 13453-350 13453-198 13453-230 13453-402 CA13453-412 13453-368 13453-364 CA13453-134 13453-122 13453-400 13453-366 CA13453-130 CA13453-370 13453-370 CA13453-426 13453-412 CA13453-388 CA13453-268 13453-134 CA13453-144 13453-130 CA13453-262 CA13453-390 CA13453-438 CA13453-436 CA13453-434 13450-214 13453-426 CA13453-432 CA13453-158 CA13453-276 CA13453-398 13453-144 CA13453-156 13453-268 CA13453-396 13453-388 13453-146 13453-262 CA13453-152 CA13453-394 CA13453-270 CA13453-150 CA13453-392 13453-390 CA13453-280 13453-392 13453-150 13453-270 13453-438 CA13450-214 CA13453-448 13450-568 13453-434 CA13453-168 CA13453-444 13453-436 CA13453-166 13453-156 13453-398 CA13453-200 13453-276 13453-158 CA13453-164 CA13453-286 CA13453-440 13453-432 13453-394 13453-152 13453-272 13453-396 CA13453-282 13453-280 13453-282 CA13453-218 13453-448 CA13453-334 13453-200 13453-166 CA13453-332 13453-444 13453-168 CA13450-568 13453-440 13453-286 13453-164 CA13453-108 13453-218 CA13453-226 13453-332 13453-334 CA13453-220 13453-450
Short-Term Detector Tubes, Draeger
Detectores de gas
Short-Term detector tubes for use with the accuro® Gas Detector Pump and Aerotest Breathing Air Tester.
  • Dräger Tubes are glass vials filled with a chemical reagent that reacts to a specific chemical or family of chemicals
  • A variety of tubes are available for the measurement of numerous different gases and vapors
Información para pedidos: For additional tubes, contact your Avantor sales representative.