VWR® Analog Dry Block Heaters
949319 949315 949318 949312 949310 949314 949313 949311 949317 949316
12621-116EA 41713.04 MXN
12621-106 12621-118 12621-104 12621-116 12621-114 12621-122 12621-112 12621-120 12621-110 12621-108
VWR® Analog Dry Block Heaters
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, blood urea nitrogen determinations, immunoassays, melting/boiling points, and a variey of other laboratory procedures.
  • Designed for multi-purpose use
  • Exceptional temperature uniformity and accuracy
  • Hold interchangeable, modular heating blocks
  • Heat to 150°C (302°F)
  • Convenient front-mounted analog controls
  • Now with Best-in-Class, Exclusive 5-Year Warranty
These high-wattage, constant-temperature block heaters are economical and versatile
Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.
All units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100°C (212°F) and high range knob adjusts from 75 to 150°C (167 to 302°F).
Each unit requires minimum bench space and features convenient front-mounted temperature adjustment knobs providing high and low temperature control.
Certificaciones: RoHS certified. All 120V units are UL and C-UL listed and CE marked.
Información para pedidos: All units include a 234 cm (92") detachable, three-wire cord and plug. Heaters require a VWR modular heating block for operation (sold separately). Additional 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.
Precaución: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.