VWR® Precision Liquid-In-Glass Thermometers
Proveedor: VWR
VWR®
60730-0011 607300111 607301111 607300011 60730-1111 60730-0111
89095-826CS 10504.2 MXN
89095-826 CA89095-822 CA89095-826 89095-822 CA89095-820 89095-820
VWR® Precision Liquid-In-Glass Thermometers
Termómetros Analogue Thermometers
Precision thermometers are made with permanently fused markings and fractional degrees to provide precision and ensure accuracy
  • Mercury free, contain blue organic liquid against lead-free white back glass
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries
  • Tested and inspected by our USA quality team
Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST.
Certificaciones: Traceable to NIST, individually serialized.
Información para pedidos: Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.