VWR® PFA Safety-Coated Liquid-In-Glass Thermometers
Proveedor: VWR
VWR®
607100811 60710-1011 60710-0811 60710-0211 607100211 607100611 60710-0911 60710-1411 607101011 60710-1311 60710-0311 60710-1111 60710-0611 60710-0511 60710-1211
89095-650CS 2761.84 MXN
89095-660 89095-650 CA89095-658 89095-648 89095-653 89095-642 89095-654 89095-640 89095-646 CA89095-644 89095-658 89095-644 CA89095-654 CA89095-642 89095-656
VWR® PFA Safety-Coated Liquid-In-Glass Thermometers
Termómetros Analogue Thermometers
Mercury free thermometers ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
  • PFA safety coating keeps thermometer liquid and glass encapsulated in the event of breakage
  • Contain blue organic liquid against lead-free white back glass
  • Tested and inspected by our USA quality team
  • Coated thermometers are just as accurate as uncoated thermometers, but are not recommended for timed tests as the coating can slow reaction time
Certificaciones: Traceable to NIST, individually serialized.
Información para pedidos: Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.