Standard Laboratory Presses, Carver
Proveedor: Carver
4386.1010000-115V 3853 3850 3851-0 4389.4020000-230V
53880-048EA 125226.72 MXN
29999-806 53880-048 53880-054 53880-050 29999-800
Standard Laboratory Presses, Carver
Presas
Ideal for research, compacting, molding rubber or plastic, laminating, and ceramics applications.
These 12- and 25-ton, manual, two-column hydraulic lab presses feature adjustable daylight openings and easy-to-read dual scale gauge calibrated in pounds and metric tons.
The 12-ton mini press requires 1/3 less bench space than the standard C press. Compact and portable, it is ideal for restricted bench space, glove box applications, or field work. Steel construction offers rigid precision and adjustable daylights. Two-post C and Mini C presses fit 15.2x15.2cm (6x6") heated platens. Two-post M presses fit 23x23cm (9x9") heated platens.
Información para pedidos: Electric hot plate sets may be used with these presses; contact your Avantor representative for more information.