VWR® Signature™ Reach-In Incubators
Proveedor: VWR
VWR® Signature™
1925A-2 1925A 1915A 1915A-2
35962-095EA 218018.25 MXN
35962-096 35962-063 35962-095 35962-062
VWR® Signature™ Reach-In Incubators
Incubadoras Incubadores estándar
The VWR® Signature™ Reach-In Incubators provide 30.8 and 38.6 cu.ft
  • Microprocessor temperature control
  • Over-temperature safety protection
  • Gentle air flow
  • Temperature uniformity to 0.8°C
  • Four interior electrical outlets
  • Pre-wired access panel for chart recorder
  • VWR® two-year limited parts and labor warranty
Both models are supplied with six shelves capable of supporting the weight of benchtop instruments. The air jacket design and heated outer door allows for uniform heat throughout the incubators. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of over temperature protection. The chamber floors are reinforced to support roller apparatus or shakers.
chamber capacities while minimizing the amount of floor space used.
Información para pedidos: Supplied with six shelves, four 110V interior electrical outlets, one access port and standard 120V electrical line cord.