Microbiological Incubators, BD Series, BINDER
Proveedor: Binder
9010-0179 9010-0189
47746-772EA 71013.76 MXN
47746-772 47746-788
Microbiological Incubators, BD Series, BINDER
Incubadoras Incubadores estándar
BINDER microbiological incubators are ideal for research, production and quality assurance.
These incubators feature the APT.Line™ preheating chamber for reliable temperature accuracy and uniformity in applications requiring constant, reproducible incubation conditions. They have a temperature range from 5º above ambient to 100ºC. Ventilation is adjustable by means of a rear exhaust and front ventilation slide. The disinfection cycle operates at 100°C for maximum hygiene.
Units have an inner tempered safety glass door, corrosion resistant stainless steel chamber and two chrome plated racks. They also carry a Class 3.1 independent temperature safety device with visual and audible temperature alarm.
Incubators are equipped with a microprocessor PID controller with LED display, integrated timer, and one ramp function. They also feature an RS-422 interface for use with APT-COM™ Data Control System software, compliant with GLP/GMP and FDA guideline 21 CFR Part 11.
The BD 23, BD 53, and BD 115 models are stackable.
Información para pedidos: Incubators include two chrome-plated shelves and are equipped with a Class 3.1, DIN 12880 safety device. For IQ/QQ qualification documentation and validation and calibration certificates, contact your Avantor sales representative.