Alcohol Burners, Boekel Scientific
Proveedor: Boekel Scientific
19691
17822-605EA 4108.59 MXN
17822-605
Alcohol Burners, Boekel Scientific
Calentadores Lámparas de alcohol
Burner is made of unbreakable spun aluminum and is spill- and tip-resistant
The wick holder screws tight against a rubber gasket for added leak resistance.
Información para pedidos: Burner is supplied with aluminum snuffer cap and two wicks.