SP Bel-Art MICRO-MILL® Grinding Mill, Bel-Art Products, a part of SP
Proveedor: Bel-Art
MICRO-MILL®
H37252-0000 H37250-0000 H37252-0001 H37254-0000
48975-407EA 75829.81 MXN
48975-407 48975-429 48975-418 16126-650
SP Bel-Art MICRO-MILL® Grinding Mill, Bel-Art Products, a part of SP
Molinos
Designed for rapid, high-speed grinding of small samples from 20 to 50 mL.
  • Built-in heat exchangers maintain temperature
  • Stainless steel or hard-faced blade
  • Operates only when cover is latched in plcae
  • Tubing connections compatible for circulating water or slurry
Stainless steel or hard-faced blade rotates at speeds up to 12,000rpm to successfully grind bauxite, bone, ceramics, coal, Devorda’s alloy, fertilizer, filter paper, fly ash, frozen tissue, glass, grain, kaolinites, leaves, limestone, pharmaceuticals, pigment blends, plastics, rocks, roots, shale, seeds, slags, soil, teeth, wood, etc
Temperature is maintained by means of built-in heat exchangers, and the control panel has an overload protection reset button and a pilot light. Tubing connections 6.4 mm (¼”) on the cover and housing accept tubes for circulating water or slurry. Cover, grinding assemby, and milling blade head are removable for cleaning. The stainless steel chamber is 54 mm diam x 25 mm deep (2⅛ x 1”) inside an aluminum housing , with an epoxy finish and non-skid rubber feet.
The ⅕ horsepower (150W) motor develops a torque of 2.2kg/cm (30oz./inch) at 12,000rpm. Unit is supplied with a 1.8m (6') 3-wire cord and grounded plug. Three models available.
To ensure safety and prevent sample loss, the grinder operates only when the cover is latched in place.
Certificaciones: CE marked.
Información para pedidos: Mills include 6-foot, 3-wire grounded cord and plug and one blade. Mill 48975-429 is complete with a stainless-steel blade and grinding chamber, with momentary hold-down switch. Mill 48975-407 is complete with stainless-steel blade and grinding chamber, including 0 to 5 minute electrical timer with ON position for continuous operation. Mill 48975-418 is complete with a hard, stellite-faced blade and grinding chamber and electrical timer.