Comply™ SteriGage™ Sterilization Integrators, 3M™
Proveedor: 3M
Comply™
MMM 1243A 7000002707
34010-019CS 6319.98 MXN
34010-019 CA34010-019
Comply™ SteriGage™ Sterilization Integrators, 3M™
Indicadores de esterilización
Provide a simple, accurate method of assuring that all conditions necessary for sterilization have been met during a sterilization cycle.
Steam sterilization integrator can be used in all steam sterilization processes, including vacuum, gravity, and flash sterilization. Integrates time, temperature, and steam just like live spore strips. Accurate to ±1 minute at 132 °C (270 °F). Works at all normal sterilization temperatures, eliminating multiple inventories.
Steam integrators are available with optional extender or recording card. Extender is ideal for verifying integrators without contaminating sterilized materials in large packs. Integrator plus extender is 23 cm (9³/₁₆") long. Recording card provides a permanent record of sterilizer use and work flow.
Integrators are held together with a temperature-resistant, pressure-sensitive adhesive. Information is displayed in a precise, easy-to-read manner to tell user whether to accept or reject the pack, or load immediately after the sterilization cycle.