Sterilization Indicator Tapes, Propper Manufacturing
Proveedor: Propper
26800900 26800600 26800400 26800500 26800800
58752-704CS 10067.5 MXN
58752-787 58752-704 58752-702 58752-781 20068-448
Sterilization Indicator Tapes, Propper Manufacturing
Indicadores de esterilización
Propper blue indicator tape and Strate-Line® tape provide means of identifying steam sterilized packs while ensuring their security during sterilization.
  • Distinguish between processed and unprocessed sterilization packages
  • Strong crepe-embossed, latex-free tape
  • Distinct white to black color change after exposure to steam
  • For use in securing wrapped surgical trays before, during, and after sterilization
Strate-Line® and blue indicator pressure-sensitive sterilization tape is used to distinguish between processed and unprocessed items. The white stripes turn to black after exposure to a steam sterilization cycle. Propper blue indicator tape features white stripes that turn to black after exposure to a steam sterilization cycle. These tape's strong adhesive and even coating allows for even dispensing and ease of application. Specifically formulated universal adhesive provides secure bonding with fabric and non-woven wraps, paper, metal and glass, yet provides easy removal without traces of residue.
Certificaciones: AAMI/ISO 11140-1 Type 1