Lindberg/Blue M Box Furnaces, 1100 °C, Thermo Scientific
Proveedor: Thermo Scientific
Lindberg/Blue M
BF51794C-1 BF51848C-1 BF51748A-1 BF51766A-1 BF51866A-1 BF51894C-1
30604-152EA 92373.69 MXN
30712-116 52346-266 30604-158 30620-004 30604-152 30604-160
Lindberg/Blue M Box Furnaces, 1100 °C, Thermo Scientific
Hornos Hornos
Small, reliable furnaces meet the specific needs of schools, as well as chemical and industrial laboratories
Choose from two types of digital controllers. All controllers display the actual and setpoint temperatures simultaneously. Furnaces with a programmable controller offer 16 programmable segments as well as user-defined proportional, integral, and derivative values. Both available controllers offer over-temperature protection.
Maximum temperature is 1100°C. All units are UL listed.
Constructed using Moldatherm®, Lindberg/Blue M’s unique ceramic fiber insulation and heating element composite, which helps to maintain uniform heat within the chamber. Low temperature outer shell is achieved through double shell construction. All Moldatherm box furnaces feature a safety door switch system that disconnects the heating unit from the power supply when the door is opened.
Información para pedidos: Furnaces feature a one-year parts and labor warranty. Models include a power cord with plug, except for 30604-156 and 30620-106 which require power cord and hard wiring.