Barnstead/Thermolyne Small Benchtop Muffle Furnaces, Type 1300 and Type 1400, Thermo Scientific
Proveedor: Thermo Scientific
Barnstead/Thermolyne
FB1315M FB1315M FB1310M FB1410M FB1310M FB1415M FB1415M FB1410M
30605-022EA 48097.28 MXN
CA30605-023 30605-023 30605-022 CA30605-022 CA30608-020 CA30608-021 30608-021 30608-020
Barnstead/Thermolyne Small Benchtop Muffle Furnaces, Type 1300 and Type 1400, Thermo Scientific
Hornos Hornos
Versatile and economical Barnstead/Thermolyne furnaces are specifically designed for ashing organic and inorganic samples, determining volatile and suspended solids, gravimetric analysis, heat treating of small steel parts, and ignition tests.
Reliable digital control ensures accurate temperature control. Turn the set point knob and the furnace automatically heats to selected temperature. Thermocouple break protection cuts power to the heating elements, preventing thermocouple failure and runaway condition.
Ceramic fiber insulation permits faster heat-up time while reducing energy consumption. Custom-formed refractory cement envelops the heating elements reducing the penetration of contaminants, extending the longevity of the heating elements. Insulated door with a convenient latching handle forms a useful shelf when open. Door safety switch protects operator by removing power to the heating elements upon opening the door.
UL listed. CSA certified.
Información para pedidos: Supplied with thermocouple and line cord with NEMA 5-15 plug. Replacement heating elements, thermocouples, as well as hearth plate to protect bottom of unit are available separately.