Barnstead/Thermolyne Compact Benchtop Muffle Furnaces, Type 1500, Thermo Scientific
Proveedor: Thermo Scientific
Barnstead/Thermolyne
FD1535M FD1530M FD1535M FD1530M
30631-230EA 71348.01 MXN
30631-230 CA30631-230 30631-232 CA30631-232
Barnstead/Thermolyne Compact Benchtop Muffle Furnaces, Type 1500, Thermo Scientific
Hornos Hornos
Economical, versatile furnaces have heavy-duty firebrick insulation surrounding the chamber opening for added durability
  • Single setpoint control, built-in door safety switch
  • Heating elements can be replaced individually, reducing operational cost
  • Counter-weighted insulated door swings upward, directing heat away from operator
Adjustable alarm or over-temperature protection setting can be used to protect the furnace or loaded chamber from excessive heat. Thermocouple break protection cuts power to the heating elements, preventing thermocouple failure and runaway conditions. Heating elements are embedded in custom-formed refractory cement, reducing the penetration of contaminants.
Información para pedidos: Replacement heating elements sold separately.