Thermolyne™ Premium Large Muffle Furnaces, Thermo Scientific
Proveedor: Thermo Scientific
Barnstead/Thermolyne
F6028C-80 F6038CM F6020C-80 F6028C F6020C F6028C-80 F6038CM F6010 F6020C-80 F6028C F6030CM F6020C F6010 F6018 F6030CM F6018
30616-051EA 86239.99 MXN
30609-026 CA30609-024 CA30609-026 CA30616-051 30616-074 30609-024 30616-051 CA30616-154 CA30616-120 30616-120 30616-154 CA74570-024 CA30616-074 74570-024 CA30616-109 30616-109
Thermolyne™ Premium Large Muffle Furnaces, Thermo Scientific
Hornos Hornos
Furnaces for fusions, brazing, enameling, drying precipitates, and volatile solids analysis.
Heating elements are located on four sides for temperature uniformity and are enveloped in custom-formed refractory cement. Reduces the damage caused by contaminant build-up on the resistant wire. Heating elements can be replaced individually, reducing operational cost of replacing all the heating elements. Heating chamber is surrounded by ceramic fiber insulation for maximum energy efficiency.
Economical, easy-to-use digital control with door safety switch and sensor break protection provides accuracy, reliability, and safety. Single setpoint automatic controller features an over-temperature alarm. Programmable model features microprocessor-based, 3-mode programmer with 16 programmable segments. The 16-segment programmer comes with 4 stored programs and includes RS-232 communications port for connection to printer or computer. Both the single setpoint automatic and programmable models have 1.27cm (1/2") fluorescent digital display with °C or °F readout.
Built-in venting system removes contaminants. Drop down door serves as a handy shelf for loading and unloading. All electrical connections through conduit connectors.
CSA certified.
Información para pedidos: All units include a thermocouple. Furnaces 74570-024 and 30616-051 are supplied with cord and plug set.