Barnstead/Thermolyne Vari-Mix™ Platform Mixer, Thermo Scientific
Proveedor: Thermo Scientific
Vari-Mix™
M48725Q M71015Q
58810-367EA 21845.84 MXN
57020-078 58810-367
Barnstead/Thermolyne Vari-Mix™ Platform Mixer, Thermo Scientific
Agitadores de laboratorio Agitadores Basculantes y de Rotación
Obtain uniform specimen suspension with the smooth rocking action of Thermo Scientific™ Vari-Mix™ and Speci-Mix™ Test Tube Rockers
Compact and precision-controlled platform mixers, all models hold 10 to 30mm diameter tubes and are ideal for blood collection tubes.