VWR® 11 mm Standard Opening Crimp-Top Vials
Proveedor: VWR
VWR®
VWC4012-1W VW60820-1232 VWC4012-2W 89239-004 89239-006
82028-404CS 5073.61 MXN
89239-006 89239-004 82028-404 66020-953 82028-402
VWR® 11 mm Standard Opening Crimp-Top Vials
Viales
Standard opening crimp-top vials have a maximum fill volume of 1.8 ml and are manufactured from inert, 33 expansion borosilicate glass.
  • Chemically inert
  • Flat Bottom
Vials are fully compatible with Agilent® 7673 and 1100 series autosamplers
Inserts are made of borosilicate glass or polypropylene.
Aluminum seals measure 11 mm.
Envase: Vials are provided in clear-pack trays.