Barnstead/Thermolyne Benchtop Muffle Furnaces, Type 47900 and Type 48000, Thermo Scientific
Proveedor: Thermo Scientific
Barnstead/Thermolyne
F48050 F48028 F47915 F48020-80 F47925-80 F47950 F47915 F48020-80 F47955 F47925 F48028 F47925 F47925-80 F48010 F48028-80 F48028-80 F47920-80 F47920-80 F48050 F47910 F47955 F47950 F47910 F48010
30610-099EA 57690 MXN
30610-216 CA30610-159 30609-942 CA30610-137 30609-986 CA30609-986 30610-159 CA30610-135 30610-137 30609-922 CA30609-942 CA30609-760 CA30610-216 30609-760 CA30609-927 CA30609-806 30610-099 30610-135 30609-748 CA30609-922 CA30609-748 30609-806 30609-927 CA30610-099
Barnstead/Thermolyne Benchtop Muffle Furnaces, Type 47900 and Type 48000, Thermo Scientific
Hornos Hornos
Compact benchtop muffle furnaces feature advanced electronic control
  • Open coil heating elements on both sides of chamber for fast heat with minimum temperature gradient
  • Ceramic insulation provides maximum energy efficiency
  • Spring-loaded ceramic-fiber door minimizes heat loss
  • Safety door interlock switch disconnects power to the heating elements whenever the door is open
  • Built-in venting system removes contaminants and moisture
CSA certified.
Pre-drilled 9.5mm (3/8") diameter port for injection of special atmospheres or independent monitoring of chamber temperature. Maximum temperature for all models is 1200°C (2192°F). Choice of four controllers: 8- or 16-segment programmable microprocessor, single setpoint (all offer over-temperature protection and digital readout), or economy digital control with sensor break protection only.
Información para pedidos: All furnace models come complete with an on/off switch, thermocouple, and grounded three-wire cord with plug. Type 48000 furnaces include a ceramic shelf.