VWR® Mini Blot Mixer
Proveedor: Avantor
VWR®
S0600-VWR
95057-436EA 10718.57 MXN
95057-436
VWR® Mini Blot Mixer
Agitadores de laboratorio Agitadores Basculantes y de Rotación
The Mini Blot Mixer is designed for processing blots and staining gels.
  • Compact
  • Lightweight
  • Portable
  • Motion: 3-Dimensional/Fixed 5°
The mixer combines the motions of an orbital shaker and rocker to produce a gentle yet thorough three dimensional orbit
For temperature controlled incubations, the mixer may be used in a cold room or incubator. The housing and platform can be easily cleaned and decontaminated.
Blot boxes are available in two sizes. The smaller boxes (95057-440) are ideal for mini protein gels. The larger boxes (95057-442) are sized for larger protein gels. Both boxes feature a hinged lid with positive catch for secure closing.
The pitch and speed are fixed to ensure the optimal conditions for processing western blots and staining gels. The tilt angle is large enough to thoroughly distribute solutions, yet small enough to prevent blot centers from drying out. As a result, minimal solution amounts can be used, conserving valuable probes and antibodies. A non-slip rubber mat is included to keep items in place while on the platform.