Barnstead/Thermolyne Multi-Tube Rotator, Thermo Scientific
Proveedor: Thermo Scientific
Barnstead/Thermolyne
4632Q
52434-650EA 17647.68 MXN
52434-650
Barnstead/Thermolyne Multi-Tube Rotator, Thermo Scientific
Agitadores de laboratorio Agitadores Basculantes y de Rotación
Ideal for use in hematology, blood-chemistry, and other applications where a continuous, gentle rocking motion is necessary.
Features bi-directional rotation. Provides a circular motion and a rocking pattern to keep liquids and solutions mixed without the potential for shearing or other damage. Rotator's four rubber feet provide stability. Double-sided, non-skid platform surface is made of white silicone and can hold 7mL, 10mL, or 15mL tubes firmly in place. Lightweight metal body is compact and is easy to transport. Angle of rotation is 20° from the horizontal plane.
Note: This product is intended for indoor use only.