VWR® Chemical Transfer Pump, Variable Flow
Proveedor: Avantor
VWR®
3388
23609-170EA 11525.26 MXN
23609-170
VWR® Chemical Transfer Pump, Variable Flow
Bombas Bombas peristálticas
Pump is designed for pulseless fluid transfer at variable flow rates.
  • Purge/Prime switch provides high-speed emptying/filling
Direct-drive engineering provides maximum motor power to the pump. Stepless “continuous sweep” variable control provides precise seamless speed control. Pumps fluids with a viscosity of up to 200 centipoise. Pump features a suction lift of 3m (10') wet, 18 cm (4") dry. Flow is easily reversible for draining tubing. The chemical-resistant wetted parts are Dupont Delrin, 304 stainless steel, Viton and Dupont Teflon. Enclosed in chemical-resistant ABS plastic case.
Certificaciones: UL listed and CSA approved.
Información para pedidos: Barbed inlet/outlet ports accept any type of tubing with an inside diameter nominal 4.8mm (3/16").