BCR® High Efficiency Mask, Berkshire
Proveedor: Berkshire
BCR®
BM.1.12
21923-584CS 12059.21 MXN
21923-584
BCR® High Efficiency Mask, Berkshire
Máscaras Máscaras para salas limpias
Three-layer mask for controlled environments.
  • Non-shedding
  • Polyurethane earloops reduce particulation
  • Headband option provides more secure facial seal
  • Ultrasonic bonding reduces contamination and outgassing
  • Cleanroom Class 1 - 100000
High efficiency (96% at 1 µm) polypropylene filter is held between 100% nylon outer layer and 100% rayon inner layer. Pressure differential of 1.98 for continuous comfort and breathability. Polyurethane ear loops can be worn around the ears or fastened with a polyethylene clasp. Ultrasonically bonded seams.
Certificaciones: ISO Class 3–8
Información para pedidos: Individually wrapped or bulk packaging options.