KIMTECH PURE® M6 Face Mask, Kimberly-Clark Professional®
Proveedor: Kimberly-Clark
Kimtech™ M6
62477 62477
10843-001CS 4766.92 MXN
CA10843-001 10843-001
KIMTECH PURE® M6 Face Mask, Kimberly-Clark Professional®
Máscaras Máscaras para salas limpias
Two-layer general-purpose mask with earloops provides comfort with BICOSOF® fabric inner facing.
  • Two layer, general purpose
  • Earloops
  • BICOSOF® fabric inner facing
Recommended for ISO Class 6 or higher cleanroom environments.
Envase: Double-bagged.