8247 R95 Respirator, 3M™
Proveedor: 3M
8247 7100160544 7000002060
56621-932CS 12152.97 MXN
CA56621-932 56621-932 CA11006-300
8247 R95 Respirator, 3M™
Respiradores Mascarillas desechables Mascarillas para partículas
This respirator provides worker protection against certain oil and non-oil based aerosols including those with nuisance levels of organic vapors such as solvents, degreasers, and resins.
  • Protects against certain oil and non-oil based aerosols
  • If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use
  • Provides minimum filtration efficiency of 95%
(Nuisance level acid gas refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use. Provides minimum filtration efficiency of 95%.
Precaución: Must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.