VWR® Disposable N95 Particulate Respirators
Proveedor: VWR
VWR®
9500-N95
89201-508CS 2822.4 MXN
89201-508
VWR® Disposable N95 Particulate Respirators
Respiradores Mascarillas desechables Mascarillas para partículas
VWR® N95 disposable respirators combine excellent safety and comfort features to ensure reliable protection and a lightweight, secure fit.
  • Meet NIOSH requirements for minimum filter efficiency of 95% against 0.3 µm particles
  • Provide protection against solid and liquid particles that do not contain oil
  • Lightweight construction allows for comfortable, extended use
An adjustable noseclip provides a secure seal against the user's face.