Survivair™ Opti-Fit™ Air-Purifying Respirators, Honeywell Safety
772000 752000 772000 758000 752000 762000 768000 778000 768000 778000 758000 762000
14216-174EA 4403.15 MXN
89013-442 89013-440 14216-170 14216-172 14216-174 89013-444
Survivair™ Opti-Fit™ Air-Purifying Respirators, Honeywell Safety
Full-face, economical respirators provide optimum fit, comfort, and superior optics
Features include durable silicone skirt, five-strap suspension, nosecup to prevent fogging, replaceable distortion-free lens with anti-scratch coating, lightweight construction for reduced head and neck stress, and ease of maintenance. Respirators accept either threaded (S-Series) or bayonet (T-Series) type cartridges. For S-Series cartridges and filters, see 13501-648 series. For T-Series cartridges and filters, see 89013-494 series.