Survivair™ ValuAir® Plus Resusable Half Mask Respirator, Honeywell Safety
Proveedor: Honeywell Safety
Survivair™ ValuAir®
303000 301000 303500 301500 302110 301500 302110 303500
89013-372EA 1167.23 MXN
CA89013-378 89013-364 89013-384 89013-372 CA89013-372 89013-358 CA89013-384 CA89013-364
Survivair™ ValuAir® Plus Resusable Half Mask Respirator, Honeywell Safety
Respiradores Mascarillas reutilizables Mascarillas
These air-purifying respirators are appropriate for use with interchangeable cartridges and particulate filters certified to NIOSH 42 CFR 84 regulations
Respirators feature an economical thermoplastic elastomer facepiece with low breathing resistance. Easy to don with no loose straps to block vision. Respirators are equipped with a soft sealing flange with rolled edges, perspiration port, and three-piece sliding suspension for comfort and fit. Forward-angled nosepiece allows for eyewear compatibility. Probed assembly respirators are equipped with a front opening for fit testing. For S-Series cartridges and filters, see 13501-648 series. For T-Series cartridges and filters, see 89013-494 series.
Half-mask respirators accommodate either Survivair™ threaded (S-Series) or bayonet (T-Series) type cartridges.