7500 Series Reusable Half Mask Respirators, 3M™
Proveedor: 3M
7100134949 7000002162 7100134948
43100-108CS 8584.02 MXN
43100-108 43100-106 43100-104
7500 Series Reusable Half Mask Respirators, 3M™
Respiradores Mascarillas reutilizables Mascarillas
These durable, reusable respirators allow the user to change the filter to protect against particulates, gases, and vapors.
  • Durable, reusable
  • Made from soft, resilient silicone and heat-resistant plastics
  • Cool Flow® exhalation valve reduces heat and moisture buildup
The respirators are made from soft, resilient silicone and heat-resistant plastics for unsurpassed comfort
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
The Cool Flow® exhalation valve is designed to make breathing easier and to help reduce heat and moisture buildup in the facepiece. The head harness is designed to adjust easily and can be worn in either traditional or drop-down mode.
Información para pedidos: Respirators are supplied without cartridges. For an assortment of compatible cartridges with stacked P100 filters, see 56621-048 series.