Nunc® Caps for 96-Well Polypropylene Plates, Thermo Scientific
Proveedor: Thermo Scientific
Nunc®
276011 276000 276011 278616 276005 276002 9503230 9503233 276002 276005 276000
44200-004CS 7393.11 MXN
CA44200-006 CA12777-962 11388-574 CA44200-002 44200-004 CA44200-004 44200-006 CA44200-030 44200-002 11388-572 12777-962
Nunc® Caps for 96-Well Polypropylene Plates, Thermo Scientific
Juntas Sellos de microplaca
These caps are for use with 0.5 ml U96, 0.45 ml V96, and 1.0 ml and 2.0 ml deep well polypropylene plates.
Choice of pre-slit silicone cap, or caps made with a self-closing thermoplastic elastomer that ensures tight sealing at low temperatures and during agitation. Caps can withstand –80 °C (–112 °F), with the exception of the silicone cap, which can withstand –20 °C (–4 °F). Compatible with automated equipment. Autoclavable. Available in different colors.
Información para pedidos: Cap 44200-002 is packed one per sleeve. Other caps are packed five per sleeve.