VWR® Films for ELISA and General Incubation, Polyester
Proveedor: VWR
VWR®
SPEVWR100 SPESVWR100 ACVWR100
60941-120CS 2138.25 MXN
60941-120 60941-064 60941-062
VWR® Films for ELISA and General Incubation, Polyester
Juntas Sellos de microplaca
These 77.5 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and well-to-well contamination, and eliminating 'edge effects' in sensitive ELISA assays.
  • Functional temperature range from −40 to +120 °C
  • Available sterile and non sterile
  • Non pierceable
Polyester films can also be used with cell culture plates for short-term storage, incubation, and containment of biohazards.
Each film L×D: 146,1×79,4 mm