PVC Film Wrap, T FRANK MCCALLS INC
Proveedor: T. Frank McCall's
56904SEAL12 56904SEAL18 44104SEL3000
52857-867EA 1996.72 MXN
52857-903 52857-867 52857-889
PVC Film Wrap, T FRANK MCCALLS INC
Juntas Películas de sellado
All-purpose, clear, polyvinyl wrap
Self-sealing. Adheres readily to glass vessels such as test tubes, flasks, cylinders, bottles, petri dishes, etc. Supplied in plain white carton.