Polax-2L Semi-Automatic Polarimeter, ATAGO®
Proveedor: Atago
RE-6721 5223-POLAX2L
47752-882EA 234488.36 MXN
47752-882 80094-062
Polax-2L Semi-Automatic Polarimeter, ATAGO®
Polarímetros
This semi-automatic polarimeter features an easy-to-read digital screen that displays angle of rotation and international sugar scale measurements.
  • Unit is suitable for applications in a variety of industries, such as pharmaceutical, perfume, cosmetic, industrial, chemical, and food manufacturing
  • Ideal for the analysis of optically active matter, including saccharin, asorbic acid, sodium glutamate, and testosterone
  • Push-button interface allows user to easily modify rotation speed
  • Long-lasting LED light source with interference filter provides a wavelength of 589 nm
Angle of rotation measurement: –179.95 to +180.00°
Angle of rotation accuracy: ±0.10°
Angle of rotation resolution: 0.05°