Outlet Oil Mist Filters, Edwards
Proveedor: Edwards
A46229000 A46220000 A46226000 A46229000 A46226000 A46220000
EVA462-26-000EA 13813.18 MXN
EVA462-29-000 CAA462-20-000 CAA462-26-000 CAA462-29-000 EVA462-20-000 EVA462-26-000
Outlet Oil Mist Filters, Edwards
Filtros Filtros de gas
For efficient removal of oil vapors/odors from vacuum pump exhaust.
Información para pedidos: Complete with NW10 or NW25 adapters, NW clamp, and centering ring for easy installation.