Easy operation, a long battery life, and reliable performance make these compact scales an excellent choice for light production, field, home, or office use
- Tare range to capacity by subtraction
Applications can include forensic testing, quality control, formulation, soil sampling, postal weighing, and dietary needs. Scales are designed with superior RF protection to deliver accurate and stable results when used in production areas or near cell phones. The large weighing platform can adequately handle large samples, and is removable for easy cleaning. Rugged plastic construction and battery powered operation allow for reliable, portable use. Standard features include a stainless steel pan, low battery indicator, overload/underload indicators, and an energy-saving auto shut-off that initiates after five minutes without activity. The low profile design includes a high contrast LCD and tactile feedback keypad.
Información para pedidos: Manufacturer's one-year warranty on parts and labor. Supplied with three AA batteries and an AC adapter. Scales 82030-832, 13500-974, and 46620-092 are supplied with a U.S. postal chart.