Scales are ideal for food portioning and checkweighing
- Tare range full capacity by subtraction
The scales can be set to act as a dedicated, low profile checkweighers for food portioning and grading. For portioning and prepackaging, the positive checkweighing mode can be used; the negative checkweighing mode can be used for portioning out of the box. The zero checkweighing mode is designed for sorting and grading operations, allowing the user to check portions against preset limits. Target weights can be quickly entered using the dedicated setpoint key. Progress toward weight targets is indicated with three LEDs (over, acceptable, and under) and a selectable audible buzzer.
Meets protection class IP 43. UL and C-UL listed. Scales are NSF certified, USDA-AMS accepted, and HACCP compliant. EC/OIML Type approved. Foot tare switch is legal for trade.
Scales are designed for use in restaurants, bakeries, food manufacturing, and other applications requiring flexible and quick operation. Scales feature large rubber pan supports; dishwasher-safe, stainless steel weighing pans; and all-stainless steel housings for sanitary cleaning and durability. A 120-hour internal rechargeable battery allows for easy portability. Large, backlit LCD displays weight in pounds up to three decimal places, as well as fractional or decimal ounces. With the push of a button, weight may also be displayed in a lb./oz. format, or for metric use, scale can display results in kilograms or grams. Mode and weighing units can be locked-in using either software or hardware locks. Software menu allows the user to recalibrate the scale using either span (two-point) or linear (three-point) calibration. Software and hardware lockouts can be used to prevent accidental recalibration.