Nalgene® Vacuum Pumps, Hand-Operated, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6131-0020 6131-0010
54903-819CS 25777.82 MXN
54903-819 54903-808
Nalgene® Vacuum Pumps, Hand-Operated, Thermo Scientific
Bombas Bombas de vació
These durable pumps easily and quickly attain and hold vacuum of 635Torr (25" Hg)
Smaller size has a pumping rate of 15mL/stroke and 3psi positive pressure at the exhaust port. Larger sizes have 36mL/stroke and 7psi positive pressure at the exhaust port. Units have a removable safety cover on exhaust port. They feature an adjustable vacuum release with trigger which works without disconnecting pump from the line. Lightweight and self-lubricating.
Información para pedidos: Supplied with two-feet of NALGENE® 8000 clear plastic tubing.