Defender™ 3000 Bench Scales, Ohaus®
Proveedor: Ohaus
Defender®
83998112 83998116 83998114 83998111
97001-302EA 29752.8 MXN
97001-302 97001-304 97001-296 97001-298
Defender™ 3000 Bench Scales, Ohaus®
Balanzas
This economic line of bench scales is ideal for general weighing and basic counting applications in production, packaging, warehouse, inventory, and shipping and receiving areas
All scales feature a type 304 stainless steel pan, painted carbon steel frame, and aluminum IP67 load cell. The indicator features a high-impact, durable ABS housing with reversible face for wall, column, or table-top mounting. Nonslip rubber leveling feet can be adjusted to exact requirements with an externally visible leveling indicator. All scales offer weighing units in lb., oz., lb.:oz., kg, and g. Integrated functions include parts counting mode with selectable sample sizes, last sample size, and Average Piece Weight data capabilities. Scales are powered by an AC adapter or by an included internal rechargeable lead acid battery (100hr. capacity). Built-in RS232 ports are standard.
The simple yet durable series is equipped with a variety of features to ensure dependable and versatile laboratory and general-purpose use.