SP Bel-Art Mini-Sieve™ Micro Sieve Set, Bel-Art Products, a part of SP
Proveedor: Bel-Art
Mini-Sieve™
F37845-1000 F37846-0000
57333-965CS 56989.55 MXN
57333-965 13197-260
SP Bel-Art Mini-Sieve™ Micro Sieve Set, Bel-Art Products, a part of SP
Tamices Tamices de ensayo
This micro sieve set consists of a stack of four interchangeable screen holding sections, a catch pan, a cover, and four screen retaining rings.
  • Interchangeable screen holding sections
  • Adjustable sorting parameters
  • Molded polypropylene sieve components
All parts are molded of polypropylene
Sorting parameters can be changed by exchanging the removable phosphor-bronze mesh inserts, (eight included, one each of 25, 35, 45, 60, 80, 120, 170, and 230 standard mesh). Each holding section is 56 mm (2¼”) I.D. x 41 mm (1⅝”) high, and holds about 60 mL.
Dimensions of the assembled set with pan and cover are 21.6 cm (8½”) high x 7.2 cm (2⅛”) maximum diameter.
Sections are friction-fit and easy to assemble and take apart for sample retrieval.
Información para pedidos: Additional mesh inserts are available seperately. They include a pack of eight Phosphor-Bronze screen inserts, one each of 25, 35, 45, 60, 80, 120, 170, and 230 standard mesh.