LiChroCART® Stainless Steel LiChrospher® Cartridges, MilliporeSigma
Proveedor: MilliporeSigma
LiChroCART® LiChrospher®
1.50843.0001 1.50958.0001 1.50159.0001 1.50824.0001 1.50826.0001 1.50834.0001 1.50638.0001 1.50600.1003 1.50958.0001 1.50154.1003 1.50636.0001 1.50833.1003 1.50829.0001 1.50840.0001 1.50963.0001 1.50638.0001 1.50641.1003 1.50742.0001 1.50833.0001 1.50928.0001 1.50601.1003 1.50823.1003 1.50837.0001 1.50961.0001 1.50826.0001 1.50155.0001 1.50742.0001 1.50834.0001 1.50640.1003 1.50635.0001 1.50983.0001 1.50155.0001 1.50833.0001 1.50159.1003 1.50984.0001 1.50937.0001 1.50842.0001 1.50943.0001 1.50158.1003 1.50987.0001 1.50987.0001 1.51908.0001 1.50154.0001 1.50828.0001 1.50602.1003 1.50931.0001 1.50937.0001 1.50847.0001 1.50602.0001 1.50600.0001 1.50931.0001 1.50963.0001 1.50155.1003 1.50839.1003 1.50148.0001 1.50641.0001 1.50984.0001 1.50603.0001 1.50154.0001 1.50601.0001 1.50843.0001 1.50600.0001 1.50639.0001 1.50928.0001 1.50943.0001 1.50605.0001 1.50641.0001 1.50847.0001 1.50822.0001 1.50602.0001 1.50601.0001 1.50642.0001 1.50840.0001 1.50858.0001 1.50642.0001 1.50603.0001 1.50148.0001 1.50158.0001 1.50983.0001 1.50822.0001 1.50961.0001 1.50158.0001 1.50639.0001 1.50837.0001 1.50829.0001 1.50636.0001 1.50635.0001 1.50858.0001 1.50981.1003 1.50605.0001
48219-456PK 38770.83 MXN
CA48219-390 48219-382 CA1.50858.0001 CA1.50605.0001 48219-388 CA48219-312 48219-588 48219-582 10811-992 CA48219-592 CA48219-394 10811-990 10811-942 48219-616 10811-940 48219-618 10811-946 10811-988 10811-944 CA48219-588 CA1.50636.0001 CA1.50928.0001 CA48219-508 10143-900 CA48219-380 10143-906 10143-904 10144-156 CA48219-388 10144-158 48219-410 48219-456 CA1.50600.0001 CA48219-582 48219-450 CA48219-382 CA1.50840.0001 48219-376 CA1.50603.0001 CA48219-616 CA1.50635.0001 10811-994 48219-404 10811-834 CA48219-456 CA1.50638.0001 10143-912 CA48219-618 10143-910 CA1.50641.0001 10143-916 CA1.50602.0001 CA48219-410 CA48219-376 48219-368 CA48219-450 48219-362 48219-442 CA48219-372 48219-562 CA1.50155.0001 10143-882 48219-514 CA48219-404 CA48219-368 CA48219-402 10811-928 10143-886 10143-128 10811-926 10143-884 CA1.50639.0001 10143-888 48219-592 48219-394 48219-390 48219-312 CA48219-562 CA48219-442 CA48219-362 10143-890 CA48219-516 48219-508 10811-934 CA48219-514 CA1.50642.0001 10811-932 CA1.50601.0001 10143-898 10812-210
LiChroCART® Stainless Steel LiChrospher® Cartridges, MilliporeSigma
Columnas de cromatografía
Cartridges are filled with reliable, versatile, traditionally produced spherical silica carrier materials.
RP-18, RP-18E (Endcapped), RP-8, and RP-8E (Endcapped) cartridges are ideal for reproducible reversed phase separation in chromatography applications including acidic, neutral, and weakly basic compounds. RP-Select B cartridges can be used to provide excellent separations of basic compounds and neutral or acidic substances. Si (silica gel) cartridges are ideal for normal phase HPLC applications. CN (cyano) cartridges are ideal for chromatography applications with complex samples with polar and hydrophobic characteristics. CN can be used as a less polar alternative to Si 60 in normal phase applications or as a less hydrophobic alternative to RP-8 in reversed phase applications. DIOL cartridges are ideal for use with complex samples with polar and hydrophobic characteristics and for exclusion chromatography applications. DIOL cartridges can be used as a less polar alternative to Si 60 in normal phase applications or as a less hydrophobic alternative (with some limitations) to RP-8 in reversed phase applications. NH₂ (amino) cartridges are ideal for reversed phase, straight phase, and ion exchange chromatography applications. Typical applications include the separation of carbohydrates (mono-, di-, and oligosaccharides) with reversed phase chromatography or the separation of nucleotides with NH₂ as a weak anion exchanger. PAH (polycyclic aromatic hydrocarbons) cartridges are highly efficient and selective HPLC cartridges designed for the high resolution separation of 16 PAH according to EPA standards 610 and 550, benzo(e)pyrene, and perylene. WP (Wide Pore) 300 RP-18 cartridges can be used to provide high-resolution separations of peptides, tRNA molecules, and low molecular weight proteins. WP 300 RP-18 cartridges enable the tailing-free separation of basic compounds and provide high recovery rates, especially in the case of highly hydrophobic peptides.
Narrow bore cartridges (2 and 3 mm Int.Ø) reduce solvent consumption and allow the handling of very small samples while achieving increased sensitivity and resolution.
Información para pedidos: All LiChroCART® cartridges require end fittings. End fittings 14229-736 are for LiChroCART® cartridges with an Int.Ø of 2, 3, 4, or 4.6 mm. End fittings 48219-720 are for LiChroCART® cartridges with an Int.Ø of 10 mm.