LiChroCART® Stainless Steel LiChrosorb® Cartridges, MilliporeSigma
Proveedor: MilliporeSigma
LiChroCART® LiChrosorb®
1.51349.0001 1.51343.0001 1.51351.0001 1.51353.0001 1.51343.0001 1.51354.0001 1.51354.0001 1.51353.0001 1.51345.0001 1.51356.0001 1.51349.0001 1.51355.0001 1.51355.0001 1.51345.0001 1.51351.0001
48219-696EA 15993.69 MXN
CA48219-696 48219-696 48219-710 CA48219-692 48219-692 CA48219-704 48219-704 CA48219-714 CA48219-700 48219-712 48219-700 CA48219-712 CA48219-710 CA48219-708 48219-708
LiChroCART® Stainless Steel LiChrosorb® Cartridges, MilliporeSigma
Columnas de cromatografía
For more than 25 years, LiChrosorb® has been one of the most successful and reliable packing materials used in HPLC.
LiChrosorb packing materials offer the complete program of nonpolar derivatives (RP-8, RP-18) and polar derivatives (Si 60).
Información para pedidos: All LiChroCART® cartridges require manuCART® end fittings. End fittings 14229-736 are for LiChroCART® cartridge columns with an Int.Ø of 4 or 4.6 mm.