Thermoplastic Ducts, Labconco
Proveedor: Labconco
4718900 5605000 5602000 4708600 7027200
30182-504EA 19954.97 MXN
30182-268 82006-918 30182-504 30182-086 30152-636
Thermoplastic Ducts, Labconco
Ducting Supplies
PVC exhaust duct is Type 1, unplasticized, schedule 40, lightweight, and corrosion-resistant
A female duct coupling is required to join two sections. Connections can be made using solvent cement. PVC is rigid and may be cut without the use of special tools